Tisk…

1-Benzylpiperidine 98%

Dodavatel: Apollo Scientific

OR916524-100G
APOSOR916524-100GEA 5290 CZK
APOSOR916524-100G
1-Benzylpiperidine 98%
1-Benzylpiperidine
Vzorec: C₁₂H₁₇N
MW: 175,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00224901
CAS číslo: 2905-56-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR