5-Methyl-3-hexanone 95%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl Isobutyl ketone, 5-Methyl-3-hexanone
OR916458-10G
OR916458-1G
OR916458-5G
APOSOR916458-10GEA
17590
CZK
APOSOR916458-10G
APOSOR916458-1G
APOSOR916458-5G
5-Methyl-3-hexanone 95%
5-Methyl-3-hexanone
Vzorec:
C₇H₁₄O MW: 114,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009319 CAS číslo: 623-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...