Methyl (S)-(-)-3-bromoisobutyrate 95%
Dodavatel: Apollo Scientific
OR916365-1G
OR916365-5G
APOSOR916365-1GEA
2480
CZK
APOSOR916365-1G
APOSOR916365-5G
Methyl (S)-(-)-3-bromoisobutyrate 95%
Methyl (S)-(-)-3-bromoisobutyrate
Vzorec:
C₅H₉BrO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00064487 CAS číslo: 98190-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...