3,4-Dihydroquinolin-1(2H)-amine 95%
Dodavatel: Apollo Scientific
OR916352-1G
OR916352-250MG
APOSOR916352-1GEA
15310
CZK
APOSOR916352-1G
APOSOR916352-250MG
3,4-Dihydroquinolin-1(2H)-amine 95%
3,4-Dihydroquinolin-1(2H)-amine
Vzorec:
C₉H₁₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD02656551 CAS číslo: 5825-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...