Tisk…

2-Methylpentyl acetate 95%

Dodavatel: Apollo Scientific

Synonyms: Acetic Acid 2-Methylpentyl Ester

OR916330-100G OR916330-25G
APOSOR916330-100GEA 9330 CZK
APOSOR916330-100G APOSOR916330-25G
2-Methylpentyl acetate 95%
2-Methylpentyl acetate
Vzorec: C₈H₁₆O₂
MW: 144.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060103
CAS číslo: 7789-99-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR