Tisk…

Tetramethylene sulphoxide 98%

Dodavatel: Apollo Scientific

Synonyms: Tetrahydrothiophene 1-oxide

OR916306-25G
APOSOR916306-25GEA 2240 CZK
APOSOR916306-25G
Tetramethylene sulphoxide 98%
Tetramethylene sulphoxide
Vzorec: C₄H₈OS
MW: 104,17 g/mol
Bod varu: 235…237 °C/760mmHg
Hustota: 1,158 g/cm³ (25 °C)
Bod vzplanutí: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00005477
CAS číslo: 1600-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR