Tisk…

3-(Benzyloxymethyl)cyclobutanone 95%

Dodavatel: Apollo Scientific

Synonyms: 3-((Benzyloxy)methyl)cyclobutanone

OR916236-1G OR916236-250MG OR916236-5G
APOSOR916236-1GEA 2030 CZK
APOSOR916236-1G APOSOR916236-250MG APOSOR916236-5G
3-(Benzyloxymethyl)cyclobutanone 95%
3-(Benzyloxymethyl)cyclobutanone
Vzorec: C₁₂H₁₄O₂
MW: 190,242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09055138
CAS číslo: 172324-67-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR