(2-Methylphenyl)methanesulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Methylbenzylsulphonyl chloride, o-Tolylmethanesulphonyl chloride
OR9162-1G
OR9162-250MG
OR9162-100MG
APOSOR9162-1GEA
0
CZK
APOSOR9162-1G
APOSOR9162-250MG
APOSOR9162-100MG
(2-Methylphenyl)methanesulphonyl chloride 97%
(2-Methylphenyl)methanesulphonyl chloride
Vzorec:
C₈H₉ClO₂S MW: 204,673 g/mol Bod tání: 51,5…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683108 CAS číslo: 92614-55-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...