2-Phenoxybenzoyl chloride 90%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR9152-1G
OR9152-250MG
APOSOR9152-1GEA
8700
CZK
APOSOR9152-1G
APOSOR9152-250MG
2-Phenoxybenzoyl chloride 90%
2-Phenoxybenzoyl chloride
Vzorec:
C₁₃H₉ClO₂ Bod tání: 37…39 °C Storage Temperature: Ambient |
MDL Number:
MFCD03424711 CAS číslo: 40501-36-8 UN: 3261 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...