Tisk…

5-Methyl-2-phenyl-1,3-oxazole-4-carbonyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR9150-100MG OR9150-1G OR9150-250MG OR9150-5G OR9150-10G
APOSOR9150-100MGEA 2410 CZK
APOSOR9150-100MG APOSOR9150-1G APOSOR9150-250MG APOSOR9150-5G APOSOR9150-10G
5-Methyl-2-phenyl-1,3-oxazole-4-carbonyl chloride 97%
5-Methyl-2-phenyl-1,3-oxazole-4-carbonyl chloride
Vzorec: C₁₁H₈ClNO₂
Bod tání: 133…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08271880
CAS číslo: 51655-71-1
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR