Tisk…

2-Methylpyrrolidine hydrochloride 95%

Dodavatel: Apollo Scientific

OR914558-1G OR914558-5G
APOSOR914558-1GEA 3020 CZK
APOSOR914558-1G APOSOR914558-5G
2-Methylpyrrolidine hydrochloride 95%
2-Methylpyrrolidine hydrochloride
Vzorec: C₅H₁₁N
MW: 121,61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08274961
CAS číslo: 54677-53-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR