2,4-Dichloro-5-methoxyphenol 97%
Dodavatel: Apollo Scientific
OR914535-1G
OR914535-250MG
APOSOR914535-1GEA
14880
CZK
APOSOR914535-1G
APOSOR914535-250MG
2,4-Dichloro-5-methoxyphenol 97%
2,4-Dichloro-5-methoxyphenol
Vzorec:
C₇H₆Cl₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD10699699 CAS číslo: 18113-13-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...