Tisk…

4'-(Dimethylamino)acetophenone 98%

Dodavatel: Apollo Scientific

Synonyms: 1-(4-(Dimethylamino)phenyl)ethanone

OR914527-100G OR914527-25G
APOSOR914527-100GEA 1990 CZK
APOSOR914527-100G APOSOR914527-25G
4'-(Dimethylamino)acetophenone 98%
4'-(Dimethylamino)acetophenone
Vzorec: C₁₀H₁₃NO
MW: 163,2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017246
CAS číslo: 2124-31-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR