Tisk…

3-Bromo-6-chloropyridazine 98%

Dodavatel: Apollo Scientific

OR914493-1G OR914493-5G
APOSOR914493-1GEA 3110 CZK
APOSOR914493-1G APOSOR914493-5G
3-Bromo-6-chloropyridazine 98%
3-Bromo-6-chloropyridazine
Vzorec: C₄H₂BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD09834814
CAS číslo: 89089-18-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR