Tisk…

3'-Chloro-o-formotoluidide 95%

Dodavatel: Apollo Scientific

OR914491-1G OR914491-5G
APOSOR914491-1GEA 7670 CZK
APOSOR914491-1G APOSOR914491-5G
3'-Chloro-o-formotoluidide 95%
3'-Chloro-o-formotoluidide
Vzorec: C₈H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD00021009
CAS číslo: 71862-02-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR