N-Boc-2-bromobenzylamine 97%
Dodavatel: Apollo Scientific
Synonyms:
N-Boc-2-bromobenzylamine, 2-Bromo-N-(tert-butoxycarbonyl)benzylamine, tert-Butyl (2-Bromobenzyl)carbamate, (2-Bromobenzyl)carbamic acid tert-Butyl ester
OR914478-5G
APOSOR914478-5GEA
2320
CZK
APOSOR914478-5G
N-Boc-2-bromobenzylamine 97%
N-Boc-2-bromobenzylamine
Vzorec:
C₁₂H₁₆BrNO₂ MW: 286,169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05664385 CAS číslo: 162356-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...