4-Phenylmorpholine 98%
Dodavatel: Apollo Scientific
Synonyms:
N-Phenylmorpholine
OR914413-100G
OR914413-1KG
OR914413-500G
APOSOR914413-100GEA
2900
CZK
APOSOR914413-100G
APOSOR914413-1KG
APOSOR914413-500G
4-Phenylmorpholine 98%
4-Phenylmorpholine
Vzorec:
C₁₀H₁₃NO MW: 163,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006166 CAS číslo: 92-53-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...