3-Benzoylbenzyl bromide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-(Bromomethyl)phenyl)(phenyl)methanone, [3-(Bromomethyl)phenyl]-phenylmethanone
OR914410-25G
OR914410-5G
APOSOR914410-25GEA
8670
CZK
APOSOR914410-25G
APOSOR914410-5G
3-Benzoylbenzyl bromide
3-Benzoylbenzyl bromide
Vzorec:
C₁₄H₁₁BrO MW: 275.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00044727 CAS číslo: 22071-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...