Tisk…

4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers) 98%

Dodavatel: Apollo Scientific

Synonyms: 1,2-Propyleneglycol sulphite, 1,2-Propanediol Cyclic Sulphite, 1,2-Propylene sulphite

OR914378-100G OR914378-25G OR914378-500G
APOSOR914378-100GEA 6220 CZK
APOSOR914378-100G APOSOR914378-25G APOSOR914378-500G
4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers) 98%
4-Methyl-1,3,2-dioxathiolane-2-oxide (mixture of isomers)
Vzorec: C₃H₆O₃S
MW: 122.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041209
CAS číslo: 1469-73-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR