3-Bromo-4-chloroaniline 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR914348-25G
OR914348-100G
OR914348-500G
APOSOR914348-25GEA
744
CZK
APOSOR914348-25G
APOSOR914348-100G
APOSOR914348-500G
3-Bromo-4-chloroaniline 98%
3-Bromo-4-chloroaniline
Vzorec:
C₆H₅BrClN Storage Temperature: Ambient |
MDL Number:
MFCD00173905 CAS číslo: 823-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...