Dimethyl-2,6-pyridinedicarboxylate 97%
Dodavatel: Apollo Scientific
Synonyms:
Dimethyl pyridine-2,6-dicarboxylate
OR914344-100G
APOSOR914344-100GEA
2570
CZK
APOSOR914344-100G
Dimethyl-2,6-pyridinedicarboxylate 97%
Dimethyl-2,6-pyridinedicarboxylate
Vzorec:
C₉H₉NO₄ MW: 195,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00134493 CAS číslo: 5453-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...