Tisk…

5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Methyl-2-phenyloxazole-4-carboxylic acid

OR9143-1G OR9143-250MG
APOSOR9143-1GEA 2240 CZK
APOSOR9143-1G APOSOR9143-250MG
5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid 97%
5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid
Vzorec: C₁₁H₉NO₃
MW: 203,2 g/mol
Bod tání: 182…183,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00275475
CAS číslo: 18735-74-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR