2-Methylpyrimidine-4-carboxylic acid 98%
Dodavatel: Apollo Scientific
OR914215-1G
OR914215-5G
APOSOR914215-1GEA
2480
CZK
APOSOR914215-1G
APOSOR914215-5G
2-Methylpyrimidine-4-carboxylic acid 98%
2-Methylpyrimidine-4-carboxylic acid
Vzorec:
C₆H₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09834807 CAS číslo: 13627-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...