2-Bromo-5-methylbenzaldehyde 98%
Dodavatel: Apollo Scientific
OR914202-5G
APOSOR914202-5GEA
1580
CZK
APOSOR914202-5G
2-Bromo-5-methylbenzaldehyde 98%
2-Bromo-5-methylbenzaldehyde
Vzorec:
C₈H₇BrO Storage Temperature: Ambient |
MDL Number:
MFCD08669550 CAS číslo: 90221-55-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...