Tisk…

2,3-Dihydroquinolin-4(1H)-one hydrochloride 97%

Dodavatel: Apollo Scientific

Synonyms: 1,2,3,4-Tetrahydro-4-quinolinone hydrochloride

OR914157-5G
APOSOR914157-5GEA 6390 CZK
APOSOR914157-5G
2,3-Dihydroquinolin-4(1H)-one hydrochloride 97%
2,3-Dihydroquinolin-4(1H)-one hydrochloride
Vzorec: C₉H₁₀ClNO
MW: 183,6400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052533
CAS číslo: 71412-22-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR