(Tetrahydro-4H-pyran-4-ylidene)acetic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
2-(Tetrahydro-4H-pyran-4-ylidene)acetic acid
OR914027-1G
OR914027-5G
APOSOR914027-1GEA
1540
CZK
APOSOR914027-1G
APOSOR914027-5G
(Tetrahydro-4H-pyran-4-ylidene)acetic acid 98%
(Tetrahydro-4H-pyran-4-ylidene)acetic acid
Vzorec:
C₇H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD04114347 CAS číslo: 130312-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...