4'-Acetoxyacetanilide 98%
Dodavatel: Apollo Scientific
Synonyms:
4-Acetamidophenyl acetate, Acetaminophen Acetate (Acetaminophen Impurity)
OR914021-25G
APOSOR914021-25GEA
4700
CZK
APOSOR914021-25G
4'-Acetoxyacetanilide 98%
4'-Acetoxyacetanilide
Vzorec:
C₁₀H₁₁NO₃ MW: 193,1992 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059205 CAS číslo: 2623-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...