(6-Methylpyridin-2-yl)methanamine 98%
Dodavatel: Apollo Scientific
OR913571-5G
APOSOR913571-5GEA
4860
CZK
APOSOR913571-5G
(6-Methylpyridin-2-yl)methanamine 98%
(6-Methylpyridin-2-yl)methanamine
Vzorec:
C₇H₁₀N₂ MW: 122,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05864530 CAS číslo: 6627-60-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...