Tisk…

3-Acetyl-6-methoxyindole 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(6-Methoxy-1H-indol-3-yl)ethanone

OR913296-1G OR913296-250MG
APOSOR913296-1GEA 3970 CZK
APOSOR913296-1G APOSOR913296-250MG
3-Acetyl-6-methoxyindole 95%
3-Acetyl-6-methoxyindole
Vzorec: C11H11NO2
MW: 189,2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08062615
CAS číslo: 99532-52-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR