Tisk…

Gabexate mesylate 98%

Dodavatel: Apollo Scientific

OR913282-1G OR913282-5G
APOSOR913282-1GEA 2150 CZK
APOSOR913282-1G APOSOR913282-5G
Gabexate mesylate 98%
Gabexate mesylate
Vzorec: C₁₆H₂₃N₃O₄·CH₃SO₃H
Bod tání: 91 °C
Storage Temperature: Freezer
MDL Number: MFCD00210299
CAS číslo: 56974-61-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR