6-Iodopyridin-2-ylamine 95%
Dodavatel: Apollo Scientific
Synonyms:
6-Iodopyridin-2-amine
OR913273-1G
OR913273-250MG
OR913273-5G
APOSOR913273-1GEA
3230
CZK
APOSOR913273-1G
APOSOR913273-250MG
APOSOR913273-5G
6-Iodopyridin-2-ylamine 95%
6-Iodopyridin-2-ylamine
Vzorec:
C₅H₅IN₂ MW: 220,01 g/mol Bod tání: 109…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD07779511 CAS číslo: 88511-25-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...