4-Chloro-1H-indole-3-carboxylic acid 95%
Dodavatel: Apollo Scientific
OR913259-1G
APOSOR913259-1GEA
2900
CZK
APOSOR913259-1G
4-Chloro-1H-indole-3-carboxylic acid 95%
4-Chloro-1H-indole-3-carboxylic acid
Vzorec:
C₉H₆ClNO₂ MW: 195,61 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07776617 CAS číslo: 23872-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...