6-Chloro-1H-indole-3-carboxylic acid 98%
Dodavatel: Apollo Scientific
OR913258-10G
OR913258-1G
OR913258-5G
APOSOR913258-10GEA
4040
CZK
APOSOR913258-10G
APOSOR913258-1G
APOSOR913258-5G
6-Chloro-1H-indole-3-carboxylic acid 98%
6-Chloro-1H-indole-3-carboxylic acid
Vzorec:
C₉H₆ClNO₂ MW: 195,6000 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07776607 CAS číslo: 766557-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...