Tisk…

4-(2-Thiazolyl)phenol 95%

Dodavatel: Apollo Scientific

OR913227-1G OR913227-5G
APOSOR913227-1GEA 2320 CZK
APOSOR913227-1G APOSOR913227-5G
4-(2-Thiazolyl)phenol 95%
4-(2-Thiazolyl)phenol
Vzorec: C₉H₇NOS
MW: 177,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368615
CAS číslo: 81015-49-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR