Tisk…

3-Methyl-5-nitro-1H-indazole 95%

Dodavatel: Apollo Scientific
Varování

OR913214-1G OR913214-250MG
APOSOR913214-1GEA 705 CZK
APOSOR913214-1G APOSOR913214-250MG
3-Methyl-5-nitro-1H-indazole 95%
3-Methyl-5-nitro-1H-indazole
Vzorec: C₈H₇N₃O₂
MW: 177,1601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07082699
CAS číslo: 40621-84-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR