Tisk…

3-(Pyrimidin-5-yl)benzaldehyde 96%

Dodavatel: Apollo Scientific

OR913126-1G OR913126-5G
APOSOR913126-1GEA 3640 CZK
APOSOR913126-1G APOSOR913126-5G
3-(Pyrimidin-5-yl)benzaldehyde 96%
3-(Pyrimidin-5-yl)benzaldehyde
Vzorec: C11H8N2O
MW: 184,1900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864833
CAS číslo: 640769-70-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR