Tisk…

7-Chloro-1H-indazole-3-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR913076-1G OR913076-250MG
APOSOR913076-1GEA 7050 CZK
APOSOR913076-1G APOSOR913076-250MG
7-Chloro-1H-indazole-3-carboxylic acid 95%
7-Chloro-1H-indazole-3-carboxylic acid
Vzorec: C₈H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD05663990
CAS číslo: 129295-32-5

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR