Tisk…

(2,3-Dichlorobenzyl)methylamine 95%

Dodavatel: Apollo Scientific

OR913034-1G OR913034-5G
APOSOR913034-1GEA 1080 CZK
APOSOR913034-1G APOSOR913034-5G
(2,3-Dichlorobenzyl)methylamine 95%
(2,3-Dichlorobenzyl)methylamine
Vzorec: C₈H₉Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD04632069
CAS číslo: 731827-07-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR