Tisk…

N-Methyl-1-(thiophen-2-yl)methanamine 95%

Dodavatel: Apollo Scientific

OR913033-1G OR913033-5G
APOSOR913033-1GEA 1950 CZK
APOSOR913033-1G APOSOR913033-5G
N-Methyl-1-(thiophen-2-yl)methanamine 95%
N-Methyl-1-(thiophen-2-yl)methanamine
Vzorec: C₆H₉NS
MW: 127,2100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04625435
CAS číslo: 58255-18-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR