Tisk…

Methyl piperidin-4-ylacetate 96%

Dodavatel: Apollo Scientific
Varování

OR913006-1G OR913006-25G OR913006-5G
APOSOR913006-1GEA 3110 CZK
APOSOR913006-1G APOSOR913006-25G APOSOR913006-5G
Methyl piperidin-4-ylacetate 96%
Methyl piperidin-4-ylacetate
Vzorec: C₈H₁₅NO₂
MW: 157,21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115353
CAS číslo: 168986-49-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR