Tisk…

3-Aminobutan-1-ol 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Amino-butan-1-ol

OR912855-10G OR912855-1G OR912855-5G
APOSOR912855-10GEA 1990 CZK
APOSOR912855-10G APOSOR912855-1G APOSOR912855-5G
3-Aminobutan-1-ol 97%
3-Aminobutan-1-ol
Vzorec: C₄H₁₁NO
MW: 89,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412694
CAS číslo: 2867-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR