Tisk…

4-(Cyclopentylamino)benzonitrile 98%

Dodavatel: Apollo Scientific

OR912839-10G OR912839-1G OR912839-5G
APOSOR912839-10GEA 14290 CZK
APOSOR912839-10G APOSOR912839-1G APOSOR912839-5G
4-(Cyclopentylamino)benzonitrile 98%
4-(Cyclopentylamino)benzonitrile
Vzorec: C₁₂H₁₄N₂
Storage Temperature: Ambient
CAS číslo: 1019512-92-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR