Tisk…

4,6-Dichloroindole 95%

Dodavatel: Apollo Scientific

Synonyms: 4,6-Dichloro-1H-indole

OR912833-1G
APOSOR912833-1GEA 3270 CZK
APOSOR912833-1G
4,6-Dichloroindole 95%
4,6-Dichloroindole
Vzorec: C₈H₅Cl₂N
MW: 186,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094990
CAS číslo: 101495-18-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR