Tisk…

4-Pyrid-2-ylbenzoic acid 98%

Dodavatel: Apollo Scientific

OR912706-1G OR912706-5G
APOSOR912706-1GEA 1910 CZK
APOSOR912706-1G APOSOR912706-5G
4-Pyrid-2-ylbenzoic acid 98%
4-Pyrid-2-ylbenzoic acid
Vzorec: C₁₂H₉NO₂
Bod tání: 237 °C
Storage Temperature: Ambient
MDL Number: MFCD01830093
CAS číslo: 4385-62-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR