(S)-2-Hydroxybutanoic acid 96%
Dodavatel: Apollo Scientific
OR912658-10G
OR912658-1G
OR912658-5G
APOSOR912658-10GEA
10520
CZK
APOSOR912658-10G
APOSOR912658-1G
APOSOR912658-5G
(S)-2-Hydroxybutanoic acid 96%
(S)-2-Hydroxybutanoic acid
Vzorec:
C₄H₈O₃ Bod tání: 50…54 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01318568 CAS číslo: 3347-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...