Ethyl-6-methylsalicylate 97%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 2-Hydroxy-6-methylbenzoate, 2-Hydroxy-6-methylbenzoic Acid Ethyl Ester, 6-Methylsalicylic Acid Ethyl Ester
OR912635-25G
APOSOR912635-25GEA
3610
CZK
APOSOR912635-25G
Ethyl-6-methylsalicylate 97%
Ethyl-6-methylsalicylate
Vzorec:
C₁₀H₁₂O₃ MW: 180,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01194299 CAS číslo: 6555-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...