Tisk…

Cinnolin-4-ol 98%

Dodavatel: Apollo Scientific

OR912607-1G OR912607-5G
APOSOR912607-1GEA 0 CZK
APOSOR912607-1G APOSOR912607-5G
Cinnolin-4-ol 98%
Cinnolin-4-ol
Vzorec: C₈H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD00956158
CAS číslo: 875-66-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR