Tisk…

(3,4-Dimethoxybenzyl)methylamine 97%

Dodavatel: Apollo Scientific

OR912588-25G OR912588-5G
APOSOR912588-25GEA 6390 CZK
APOSOR912588-25G APOSOR912588-5G
(3,4-Dimethoxybenzyl)methylamine 97%
(3,4-Dimethoxybenzyl)methylamine
Vzorec: C₁₀H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00837593
CAS číslo: 63-64-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR