Tisk…

6-Hydroxy-1,2,3,4-tetrahydroquinoline 95%

Dodavatel: Apollo Scientific

Synonyms: 1,2,3,4-Tetrahydroquinolin-6-ol

OR912575-1G
APOSOR912575-1GEA 0 CZK
APOSOR912575-1G
6-Hydroxy-1,2,3,4-tetrahydroquinoline 95%
6-Hydroxy-1,2,3,4-tetrahydroquinoline
Vzorec: C₉H₁₁NO
MW: 149,1897 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00800243
CAS číslo: 3373-00-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR