Tisk…

N-Boc-aniline 98%

Dodavatel: Apollo Scientific

Synonyms: tert-Butyl phenylcarbamate

OR912520-100G OR912520-25G
APOSOR912520-100GEA 6750 CZK
APOSOR912520-100G APOSOR912520-25G
N-Boc-aniline 98%
N-Boc-aniline
Vzorec: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00043723
CAS číslo: 3422-01-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR